Methyl (1R,2S,3S,4S,6R)-5-oxotricyclo[2.2.1.02,6]heptane-3-carboxylate
Dodavatel: Apollo Scientific
Synonyms:
Tricyclo[2.2.1.02,6]heptane-3-carboxylic acid, 5-oxo-, methyl ester, stereoisomer
OR470725-1G
OR470725-5G
APOSOR470725-1GEA
6980
CZK
APOSOR470725-1G
APOSOR470725-5G
Methyl (1R,2S,3S,4S,6R)-5-oxotricyclo[2.2.1.02,6]heptane-3-carboxylate
Methyl (1R,2S,3S,4S,6R)-5-oxotricyclo[2.2.1.02,6]heptane-3-carboxylate
Vzorec:
C₉H₁₀O₃ MW: 166,174 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28096495 CAS číslo: 79356-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...