Tisk…

1-[2-[(Tetrahydropyran-4-yl)oxy]phenyl]methylamine

Dodavatel: Apollo Scientific

OR470699-1G OR470699-250MG OR470699-5G
APOSOR470699-1GEA 17130 CZK
APOSOR470699-1G APOSOR470699-250MG APOSOR470699-5G
1-[2-[(Tetrahydropyran-4-yl)oxy]phenyl]methylamine
1-[2-[(Tetrahydropyran-4-yl)oxy]phenyl]methylamine
Vzorec: C₁₂H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD09025871
CAS číslo: 898289-33-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR