Tisk…

1,3-Bis[4-(tert-butyl)phenyl]-2,2-Bis[(S)-4-phenyl-4,5-dihydro-2-oxazolyl]propane

Dodavatel: Apollo Scientific
Varování

Synonyms: (4S,4'S)-2,2'-(1,3-Bis[4-(t-butyl)phenyl)propane-2,2-diyl]bis(4-phenyl-4,5-dihydrooxazole)

OR470682-1G OR470682-250MG
APOSOR470682-1GEA 11210 CZK
APOSOR470682-1G APOSOR470682-250MG
1,3-Bis[4-(tert-butyl)phenyl]-2,2-Bis[(S)-4-phenyl-4,5-dihydro-2-oxazolyl]propane
1,3-Bis[4-(tert-butyl)phenyl]-2,2-Bis[(S)-4-phenyl-4,5-dihydro-2-oxazolyl]propane
Vzorec: C₄₁H₄₆N₂O₂
MW: 598.816 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26404519
CAS číslo: 1428328-51-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR