Tisk…

4-(tert-Butoxy)benzamide

Dodavatel: Apollo Scientific

OR470674-1G OR470674-5G
APOSOR470674-1GEA 11310 CZK
APOSOR470674-1G APOSOR470674-5G
4-(tert-Butoxy)benzamide
4-(tert-Butoxy)benzamide
Vzorec: C₁₁H₁₅NO₂
MW: 193,242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840534
CAS číslo: 99985-67-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR