Tisk…

4-(Dibenzylamino)butanoic acid

Dodavatel: Apollo Scientific

OR470669-5G OR470669-1G
APOSOR470669-5GEA 14220 CZK
APOSOR470669-5G APOSOR470669-1G
4-(Dibenzylamino)butanoic acid
4-(Dibenzylamino)butanoic acid
Vzorec: C₁₈H₂₁NO₂
MW: 283.365 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00889235
CAS číslo: 106518-44-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR