2,4-Dichloro-1-(2,2-dibromovinyl)benzene
Dodavatel: Apollo Scientific
OR470653-1G
OR470653-5G
APOSOR470653-1GEA
11310
CZK
APOSOR470653-1G
APOSOR470653-5G
2,4-Dichloro-1-(2,2-dibromovinyl)benzene
2,4-Dichloro-1-(2,2-dibromovinyl)benzene
Vzorec:
C₈H₄Br₂Cl₂ MW: 330,831 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24842545 CAS číslo: 212138-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...