Tisk…

3-(2-Methylphenoxy)propanenitrile

Dodavatel: Apollo Scientific

OR470652-1G OR470652-5G
APOSOR470652-1GEA 5530 CZK
APOSOR470652-1G APOSOR470652-5G
3-(2-Methylphenoxy)propanenitrile
3-(2-Methylphenoxy)propanenitrile
Vzorec: C₁₀H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD03150781
CAS číslo: 25268-05-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR