Tisk…

Ethyl 4-Bromo-1-naphthoate

Dodavatel: Apollo Scientific

OR470636-25G OR470636-5G
APOSOR470636-25GEA 31910 CZK
APOSOR470636-25G APOSOR470636-5G
Ethyl 4-Bromo-1-naphthoate
Ethyl 4-Bromo-1-naphthoate
Vzorec: C₁₃H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD16038781
CAS číslo: 51934-43-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR