Methyl 5-Bromo-2-Methyl-2H-1,2,3-triazole-4-carboxylate
Dodavatel: Apollo Scientific
OR470618-1G
OR470618-250MG
APOSOR470618-1GEA
30720
CZK
APOSOR470618-1G
APOSOR470618-250MG
Methyl 5-Bromo-2-Methyl-2H-1,2,3-triazole-4-carboxylate
Methyl 5-Bromo-2-Methyl-2H-1,2,3-triazole-4-carboxylate
Vzorec:
C₅H₆BrN₃O₂ MW: 220,024 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23381101 CAS číslo: 1372711-70-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...