Tisk…

Methyl 5-Methoxy-2-thiophenecarboxylate

Dodavatel: Apollo Scientific

OR470576-1G OR470576-250MG OR470576-5G
APOSOR470576-1GEA 17130 CZK
APOSOR470576-1G APOSOR470576-250MG APOSOR470576-5G
Methyl 5-Methoxy-2-thiophenecarboxylate
Methyl 5-Methoxy-2-thiophenecarboxylate
Vzorec: C₇H₈O₃S
MW: 172,202 g/mol
Bod tání: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22682797
CAS číslo: 77133-25-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR