Tisk…

N,4-Dimethyl-N-[4-(trimethylsilyl)phenyl]benzenesulphonamide

Dodavatel: Apollo Scientific

OR470569-1G OR470569-5G
APOSOR470569-1GEA 5820 CZK
APOSOR470569-1G APOSOR470569-5G
N,4-Dimethyl-N-[4-(trimethylsilyl)phenyl]benzenesulphonamide
N,4-Dimethyl-N-[4-(trimethylsilyl)phenyl]benzenesulphonamide
Vzorec: C₁₇H₂₃NO₂SSi
MW: 333,521 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22682803
CAS číslo: 1251740-03-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR