3-(3-Piperidyl)-1-propanol
Dodavatel: Apollo Scientific
OR470567-5G
OR470567-1G
APOSOR470567-5GEA
66790
CZK
APOSOR470567-5G
APOSOR470567-1G
3-(3-Piperidyl)-1-propanol
3-(3-Piperidyl)-1-propanol
Vzorec:
C₈H₁₇NO Storage Temperature: Ambient |
MDL Number:
MFCD06408777 CAS číslo: 25175-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...