Tisk…

3-(tert-Butyldimethylsilyloxy)glutaric acid

Dodavatel: Apollo Scientific

OR470470-25G OR470470-5G
APOSOR470470-25GEA 28400 CZK
APOSOR470470-25G APOSOR470470-5G
3-(tert-Butyldimethylsilyloxy)glutaric acid
3-(tert-Butyldimethylsilyloxy)glutaric acid
Vzorec: C₁₁H₂₂O₅Si
Storage Temperature: Ambient
MDL Number: MFCD21641339
CAS číslo: 113794-48-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR