2-Ethyloxazolo[5,4-b]pyridine
Dodavatel: Apollo Scientific
OR470465-1G
OR470465-250MG
OR470465-5G
APOSOR470465-1GEA
17130
CZK
APOSOR470465-1G
APOSOR470465-250MG
APOSOR470465-5G
2-Ethyloxazolo[5,4-b]pyridine
2-Ethyloxazolo[5,4-b]pyridine
Vzorec:
C₈H₈N₂O MW: 148,162 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18384507 CAS číslo: 856990-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...