Tisk…

2-(2-Benzimidazolylamino)-6-methylpyrimidin-4(3H)-one

Dodavatel: Apollo Scientific

OR470446-1G OR470446-5G
APOSOR470446-1GEA 8140 CZK
APOSOR470446-1G APOSOR470446-5G
2-(2-Benzimidazolylamino)-6-methylpyrimidin-4(3H)-one
2-(2-Benzimidazolylamino)-6-methylpyrimidin-4(3H)-one
Vzorec: C₁₂H₁₁N₅O
MW: 241,249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03812593
CAS číslo: 42389-40-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR