Tisk…

α-Cyclopropyl-3-pyridinemethanol

Dodavatel: Apollo Scientific

OR470430-5G OR470430-1G
APOSOR470430-5GEA 14680 CZK
APOSOR470430-5G APOSOR470430-1G
α-Cyclopropyl-3-pyridinemethanol
α-Cyclopropyl-3-pyridinemethanol
Vzorec: C₉H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD12808142
CAS číslo: 155047-86-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR