6-(4-Formylphenyl)-2-pyridinecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR470428-1G
OR470428-250MG
APOSOR470428-1GEA
17130
CZK
APOSOR470428-1G
APOSOR470428-250MG
6-(4-Formylphenyl)-2-pyridinecarboxylic acid
6-(4-Formylphenyl)-2-pyridinecarboxylic acid
Vzorec:
C₁₃H₉NO₃ MW: 227,215 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05150364 CAS číslo: 566198-31-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...