1-Boc-4-(2-pyridyloxy)piperidine
Dodavatel: Apollo Scientific
OR470420-1G
OR470420-5G
APOSOR470420-1GEA
7280
CZK
APOSOR470420-1G
APOSOR470420-5G
1-Boc-4-(2-pyridyloxy)piperidine
1-Boc-4-(2-pyridyloxy)piperidine
Vzorec:
C₁₅H₂₂N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD09861999 CAS číslo: 313490-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...