Tisk…

3-[5-(Hydroxymethyl)-2-pyridyl]benzonitrile

Dodavatel: Apollo Scientific

OR470403-1G OR470403-250MG
APOSOR470403-1GEA 28400 CZK
APOSOR470403-1G APOSOR470403-250MG
3-[5-(Hydroxymethyl)-2-pyridyl]benzonitrile
3-[5-(Hydroxymethyl)-2-pyridyl]benzonitrile
Vzorec: C₁₃H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD06410453
CAS číslo: 887974-34-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR