5-(3-Nitrophenyl)-2-thiophenecarbaldehyde
Dodavatel: Apollo Scientific
OR470373-1G
OR470373-5G
APOSOR470373-1GEA
11610
CZK
APOSOR470373-1G
APOSOR470373-5G
5-(3-Nitrophenyl)-2-thiophenecarbaldehyde
5-(3-Nitrophenyl)-2-thiophenecarbaldehyde
Vzorec:
C₁₁H₇NO₃S MW: 233,243 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06410197 CAS číslo: 38401-73-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...