Tisk…

[6-(2-Chlorophenyl)-3-pyridyl]methanol

Dodavatel: Apollo Scientific

OR470329-1G OR470329-5G
APOSOR470329-1GEA 17390 CZK
APOSOR470329-1G APOSOR470329-5G
[6-(2-Chlorophenyl)-3-pyridyl]methanol
[6-(2-Chlorophenyl)-3-pyridyl]methanol
Vzorec: C₁₂H₁₀ClNO
MW: 219,667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410435
CAS číslo: 887974-37-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR