3-Methyl-4-(methylsulphonamido)benzoic acid
Dodavatel: Apollo Scientific
OR470323-1G
OR470323-5G
APOSOR470323-1GEA
6980
CZK
APOSOR470323-1G
APOSOR470323-5G
3-Methyl-4-(methylsulphonamido)benzoic acid
3-Methyl-4-(methylsulphonamido)benzoic acid
Vzorec:
C₉H₁₁NO₄S Storage Temperature: Ambient |
MDL Number:
MFCD09045026 CAS číslo: 892878-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...