2-Chloro-6-(methylsulphonamido)benzoic acid
Dodavatel: Apollo Scientific
OR470322-1G
OR470322-5G
APOSOR470322-1GEA
6980
CZK
APOSOR470322-1G
APOSOR470322-5G
2-Chloro-6-(methylsulphonamido)benzoic acid
2-Chloro-6-(methylsulphonamido)benzoic acid
Vzorec:
C₈H₈ClNO₄S Storage Temperature: Ambient |
MDL Number:
MFCD18872325 CAS číslo: 1314406-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...