4-Methyl-3-(methylsulphonamido)benzoic acid
Dodavatel: Apollo Scientific
OR470315-1G
OR470315-5G
APOSOR470315-1GEA
6980
CZK
APOSOR470315-1G
APOSOR470315-5G
4-Methyl-3-(methylsulphonamido)benzoic acid
4-Methyl-3-(methylsulphonamido)benzoic acid
Vzorec:
C₉H₁₁NO₄S Storage Temperature: Ambient |
MDL Number:
MFCD09049488 CAS číslo: 450368-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...