2-[(Cyclopropylcarbonyl)amino]-5-methylbenzoic acid
Dodavatel: Apollo Scientific
OR470313-1G
OR470313-5G
APOSOR470313-1GEA
6980
CZK
APOSOR470313-1G
APOSOR470313-5G
2-[(Cyclopropylcarbonyl)amino]-5-methylbenzoic acid
2-[(Cyclopropylcarbonyl)amino]-5-methylbenzoic acid
Vzorec:
C₁₂H₁₃NO₃ MW: 219,237 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09937124 CAS číslo: 1016817-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...