4-Chloro-2-[(cyclopropylcarbonyl)amino]benzoic acid
Dodavatel: Apollo Scientific
OR470309-1G
OR470309-5G
APOSOR470309-1GEA
6980
CZK
APOSOR470309-1G
APOSOR470309-5G
4-Chloro-2-[(cyclopropylcarbonyl)amino]benzoic acid
4-Chloro-2-[(cyclopropylcarbonyl)amino]benzoic acid
Vzorec:
C₁₁H₁₀ClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09049997 CAS číslo: 926242-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...