2-[(Cyclopropylcarbonyl)amino]-4,5-dimethoxybenzoic acid
Dodavatel: Apollo Scientific
OR470303-1G
OR470303-5G
APOSOR470303-1GEA
6980
CZK
APOSOR470303-1G
APOSOR470303-5G
2-[(Cyclopropylcarbonyl)amino]-4,5-dimethoxybenzoic acid
2-[(Cyclopropylcarbonyl)amino]-4,5-dimethoxybenzoic acid
Vzorec:
C₁₃H₁₅NO₅ MW: 265,262 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03081977 CAS číslo: 496913-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...