Tisk…

3-Methyl-1-(3-pyridyl)-1-butylamine

Dodavatel: Apollo Scientific

OR470279-1G OR470279-5G
APOSOR470279-1GEA 5820 CZK
APOSOR470279-1G APOSOR470279-5G
3-Methyl-1-(3-pyridyl)-1-butylamine
3-Methyl-1-(3-pyridyl)-1-butylamine
Vzorec: C₁₀H₁₆N₂
Storage Temperature: Ambient
MDL Number: MFCD09802327
CAS číslo: 938459-12-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR