3-Methyl-1-(4-pyridyl)-1-butylamine
Dodavatel: Apollo Scientific
OR470278-1G
OR470278-5G
APOSOR470278-1GEA
7280
CZK
APOSOR470278-1G
APOSOR470278-5G
3-Methyl-1-(4-pyridyl)-1-butylamine
3-Methyl-1-(4-pyridyl)-1-butylamine
Vzorec:
C₁₀H₁₆N₂ Storage Temperature: Ambient |
MDL Number:
MFCD12153948 CAS číslo: 1178634-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...