Tisk…

1-(3-Pyridyl)-1-butylamine

Dodavatel: Apollo Scientific
Varování

OR470277-1G OR470277-5G
APOSOR470277-1GEA 7280 CZK
APOSOR470277-1G APOSOR470277-5G
1-(3-Pyridyl)-1-butylamine
1-(3-Pyridyl)-1-butylamine
Vzorec: C₉H₁₄N₂
MW: 150,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01691643
CAS číslo: 90565-27-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR