2-(Boc-amino)-2-(1,2,3,4-tetrahydro-2-naphthyl)acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR470267-250MG
OR470267-1G
APOSOR470267-250MGEA
18190
CZK
APOSOR470267-250MG
APOSOR470267-1G
2-(Boc-amino)-2-(1,2,3,4-tetrahydro-2-naphthyl)acetic acid
2-(Boc-amino)-2-(1,2,3,4-tetrahydro-2-naphthyl)acetic acid
Vzorec:
C₁₇H₂₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD04115525 CAS číslo: 936214-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...