2-Methyl-1-(4-pyridyl)-1-propylamine
Dodavatel: Apollo Scientific
OR470263-1G
OR470263-5G
APOSOR470263-1GEA
7280
CZK
APOSOR470263-1G
APOSOR470263-5G
2-Methyl-1-(4-pyridyl)-1-propylamine
2-Methyl-1-(4-pyridyl)-1-propylamine
Vzorec:
C₉H₁₄N₂ MW: 150,221 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12153944 CAS číslo: 62398-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...