Tisk…

2-Methyl-1-(3-pyridyl)-1-propylamine

Dodavatel: Apollo Scientific

OR470262-1G OR470262-5G OR470262-250MG
APOSOR470262-1GEA 4200 CZK
APOSOR470262-1G APOSOR470262-5G APOSOR470262-250MG
2-Methyl-1-(3-pyridyl)-1-propylamine
2-Methyl-1-(3-pyridyl)-1-propylamine
Vzorec: C₉H₁₄N₂
MW: 150,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971213
CAS číslo: 343270-47-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR