Tisk…

1-Boc-5-bromo-6-methoxy-1H-indazole

Dodavatel: Apollo Scientific

OR470257-1G OR470257-5G
APOSOR470257-1GEA 11310 CZK
APOSOR470257-1G APOSOR470257-5G
1-Boc-5-bromo-6-methoxy-1H-indazole
1-Boc-5-bromo-6-methoxy-1H-indazole
Vzorec: C₁₃H₁₅BrN₂O₃
MW: 327,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18911745
CAS číslo: 1305320-56-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR