(S)-3-Hydroxy-N,N-dimethylpyrrolidine-1-carboxamide
Dodavatel: Apollo Scientific
|
Varování
|
OR470218-1G
OR470218-250MG
APOSOR470218-1GEA
6020
CZK
APOSOR470218-1G
APOSOR470218-250MG
(S)-3-Hydroxy-N,N-dimethylpyrrolidine-1-carboxamide
(S)-3-Hydroxy-N,N-dimethylpyrrolidine-1-carboxamide
Vzorec:
C₇H₁₄N₂O₂ MW: 158,198 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18651720 CAS číslo: 1305322-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...