(R)-3-Hydroxy-N,N-dimethylpyrrolidine-1-carboxamide
Dodavatel: Apollo Scientific
|
Varování
|
OR470217-1G
OR470217-5G
APOSOR470217-1GEA
8700
CZK
APOSOR470217-1G
APOSOR470217-5G
(R)-3-Hydroxy-N,N-dimethylpyrrolidine-1-carboxamide
(R)-3-Hydroxy-N,N-dimethylpyrrolidine-1-carboxamide
Vzorec:
C₇H₁₄N₂O₂ MW: 158,198 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18651719 CAS číslo: 1315054-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...