2-[Bis(methylsulphonyl)amino]benzonitrile
Dodavatel: Apollo Scientific
OR470206-25G
OR470206-5G
APOSOR470206-25GEA
37030
CZK
APOSOR470206-25G
APOSOR470206-5G
2-[Bis(methylsulphonyl)amino]benzonitrile
2-[Bis(methylsulphonyl)amino]benzonitrile
Vzorec:
C₉H₁₀N₂O₄S₂ MW: 274.317 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01648439 CAS číslo: 235100-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...