Tisk…

Methyl 3-(1-Pyrrolidinyl)benzoate

Dodavatel: Apollo Scientific

OR470203-1G OR470203-5G
APOSOR470203-1GEA 4930 CZK
APOSOR470203-1G APOSOR470203-5G
Methyl 3-(1-Pyrrolidinyl)benzoate
Methyl 3-(1-Pyrrolidinyl)benzoate
Vzorec: C₁₂H₁₅NO₂
MW: 205.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06740075
CAS číslo: 186086-71-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR