Tisk…

4-(Imidazol-2-yl)benzonitrile

Dodavatel: Apollo Scientific

OR470197-1G OR470197-5G
APOSOR470197-1GEA 11310 CZK
APOSOR470197-1G APOSOR470197-5G
4-(Imidazol-2-yl)benzonitrile
4-(Imidazol-2-yl)benzonitrile
Vzorec: C₁₀H₇N₃
MW: 169,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863386
CAS číslo: 98298-49-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR