Tisk…

3-(2-Imidazolyl)aniline hydrochloride

Dodavatel: Apollo Scientific

OR470194-1G OR470194-5G
APOSOR470194-1GEA 11310 CZK
APOSOR470194-1G APOSOR470194-5G
3-(2-Imidazolyl)aniline hydrochloride
3-(2-Imidazolyl)aniline hydrochloride
Vzorec: C₉H₉N₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD03206691
CAS číslo: 1261269-04-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR