Ethyl 2-(3-Methoxyphenoxy)propanoate
Dodavatel: Apollo Scientific
OR470174-25G
OR470174-5G
APOSOR470174-25GEA
28400
CZK
APOSOR470174-25G
APOSOR470174-5G
Ethyl 2-(3-Methoxyphenoxy)propanoate
Ethyl 2-(3-Methoxyphenoxy)propanoate
Vzorec:
C₁₂H₁₆O₄ MW: 224,253 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16089521 CAS číslo: 141289-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...