Methyl 2-Amino-4-(1-imidazolyl)benzoate
Dodavatel: Apollo Scientific
OR470171-1G
OR470171-5G
APOSOR470171-1GEA
5660
CZK
APOSOR470171-1G
APOSOR470171-5G
Methyl 2-Amino-4-(1-imidazolyl)benzoate
Methyl 2-Amino-4-(1-imidazolyl)benzoate
Vzorec:
C₁₁H₁₁N₃O₂ MW: 217.224 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18651685 CAS číslo: 1272756-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...