Tisk…

2-Acetyl-3,5-bis(benzyloxy)hydroquinone

Dodavatel: Apollo Scientific

OR470155-1G OR470155-5G
APOSOR470155-1GEA 12770 CZK
APOSOR470155-1G APOSOR470155-5G
2-Acetyl-3,5-bis(benzyloxy)hydroquinone
2-Acetyl-3,5-bis(benzyloxy)hydroquinone
Vzorec: C₂₂H₂₀O₅
MW: 364,391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11559033
CAS číslo: 1083181-35-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR