Tisk…

2,3,3-Trimethyl-2-butanol

Dodavatel: Apollo Scientific
Varování

OR470142-1G OR470142-1KG
APOSOR470142-1GEA 3540 CZK
APOSOR470142-1G APOSOR470142-1KG
2,3,3-Trimethyl-2-butanol
2,3,3-Trimethyl-2-butanol
Vzorec: C₇H₁₆O
MW: 116,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060887
CAS číslo: 594-83-2
UN: 1987
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR