Tisk…

Ethyl N-Boc-3-oxopiperidine-2-carboxylate

Dodavatel: Apollo Scientific

Synonyms: 1-tert-Butyl 2-ethyl 3-oxopiperidine-1,2-dicarboxylate

OR470125-1G OR470125-5G
APOSOR470125-1GEA 4200 CZK
APOSOR470125-1G APOSOR470125-5G
Ethyl N-Boc-3-oxopiperidine-2-carboxylate
Ethyl N-Boc-3-oxopiperidine-2-carboxylate
Vzorec: C₁₃H₂₁NO₅
MW: 271,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16883080
CAS číslo: 1245782-62-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR