Tisk…

(S)-N-Methyl-N-(2-propynyl)-2,3-dihydroinden-1-amine

Dodavatel: Apollo Scientific

OR470120-1G OR470120-5G
APOSOR470120-1GEA 7840 CZK
APOSOR470120-1G APOSOR470120-5G
(S)-N-Methyl-N-(2-propynyl)-2,3-dihydroinden-1-amine
(S)-N-Methyl-N-(2-propynyl)-2,3-dihydroinden-1-amine
Vzorec: C₁₃H₁₅N
MW: 185,265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367856
CAS číslo: 124192-86-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR