Tisk…

(4-Carbamoylphenoxy)acetic acid

Dodavatel: Apollo Scientific

OR470098-1G OR470098-5G
APOSOR470098-1GEA 8700 CZK
APOSOR470098-1G APOSOR470098-5G
(4-Carbamoylphenoxy)acetic acid
(4-Carbamoylphenoxy)acetic acid
Vzorec: C₉H₉NO₄
MW: 195.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02600649
CAS číslo: 29936-86-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR