Methyl 1-Boc-4-(Cbz-amino)piperidine-4-carboxylate
Dodavatel: Apollo Scientific
OR470091-1G
OR470091-5G
APOSOR470091-1GEA
14220
CZK
APOSOR470091-1G
APOSOR470091-5G
Methyl 1-Boc-4-(Cbz-amino)piperidine-4-carboxylate
Methyl 1-Boc-4-(Cbz-amino)piperidine-4-carboxylate
Vzorec:
C₂₀H₂₈N₂O₆ MW: 392,446 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16495881 CAS číslo: 392331-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...