1-Acetylpyrazolo[3,4-c]pyridine
Dodavatel: Apollo Scientific
OR470089-1G
OR470089-5G
APOSOR470089-1GEA
7280
CZK
APOSOR470089-1G
APOSOR470089-5G
1-Acetylpyrazolo[3,4-c]pyridine
1-Acetylpyrazolo[3,4-c]pyridine
Vzorec:
C₈H₇N₃O Storage Temperature: Ambient |
MDL Number:
MFCD16495877 CAS číslo: 52090-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...