Tisk…

7-Acetamido-3,4-dihydro-2H-1,5-benzodioxepine

Dodavatel: Apollo Scientific

OR470086-1G OR470086-5G
APOSOR470086-1GEA 8700 CZK
APOSOR470086-1G APOSOR470086-5G
7-Acetamido-3,4-dihydro-2H-1,5-benzodioxepine
7-Acetamido-3,4-dihydro-2H-1,5-benzodioxepine
Vzorec: C₁₁H₁₃NO₃
MW: 207.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11717845
CAS číslo: 33631-94-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR